Learn More
Brady™ V-Loc Breakaway System - U-Channel With Wedge and Adapter
Catalog No. 19000345 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Brown/White
Catalog No. Color
19-000-345 Brown/White
1 options
Catalog No. 19-000-345 Supplier Brady™ Supplier No. 78993
Add to Cart
Add to Cart
V-Loc Breakaway System - U-Channel With Wedge and Adapter allows posts to break off cleanly at ground level, resulting in less damage to vehicles.
  • Brady's V-Loc Breakaway system allows posts to break off cleanly at ground level resulting in less damage to vehicles.
  • The V-Loc Breakaway system can safely be installed in concrete, asphalt or dirt by one person in a matter of minutes.
  • Posts are easily replaced by removing the locking wedge and placing a new post in the socket.
  • Strong steel fins hold the socket securely in the ground resisting movement from wind vibration.
Compliance
MUTCD
Specifications
Product Type Post Breakaway System
Color Brown/White
Material Steel
Description Breakaway System - U-Channel With Wedge and Adapter
Diameter (English) 1.9 in.
Height (English) 30 in.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.