Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Vendsorb Oil Absorbent Rolls

Catalog No. 17111170 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
7.5 x 50 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-111-170 7.5 x 50 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-170 Supplier Brady™ Supplier No. VSO75P

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Absorbs petroleum-based oils, solvents and other non-water liquids

  • Conveniently sized
  • Floats indefinitely even when saturated with oil
  • Perforated and dimpled
  • Available in two sizes: 12 x 60 in. and 7.5 x 50 in.

Specifications

Type Oil Absorbent Roll
Packaging Quantity 20/Cs.
Dimensions (L x W) 7.5 x 50 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.