Learn More
Description
- High gloss, durable vinyl with pressure-sensitive rubber-based adhesive withstands most common chemicals, solvents, oils and water
- Encapsulated graphics are sealed beneath a protective layer of abrasion-resistant vinyl
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Designed to withstand tough indoor conditions
Specifications
Specifications
| Certifications/Compliance | OSHA 1910.44 |
| Adhesive Type | Rubber, Pressure-sensitive |
| Color | Magenta, Yellow |
| Finish | High Gloss |
| Format | Tape |
| Material | Vinyl |
| Product Type | Warning Tape |
| Sterility | Non-sterile |
| For Use With (Application) | For safety marking of obstructions, low hanging objects, pillars, passageways, aisles, any other potentially hazardous areas or objects |
| Gloss | 89 Gardner Units |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.