Learn More
Brady™ Brady Workstation Advanced Sequence Software, Minimum RAM Required 2GB, E-Media
Catalog No. 19129120 Shop All Brady Corporation Products
Change view
Click to view available options
Brady Workstation Advanced Sequence Software, Minimum RAM Required 2GB, E-Media
Catalog No.
19-129-120 Brady Workstation Advanced Sequence Software, Minimum RAM Required 2GB, E-Media
1 options
Catalog No. 19-129-120 Supplier Brady™ Supplier No. BWSASEM
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Brady™ Workstation Advanced Sequence Software allows users to define a sequence with multiple counters and create a separate label for each value in the sequence
Features:
  • Supports alpha and numeric sequences, plus hex, octal, binary, and custom sequences
  • Can include independent and dependent components
  • Drag and drop data anywhere on your label
Specifications
For Use With (Equipment) Brady Label Printers
Resolution 1024 x 768
Software Type Brady Workstation Advanced Sequence Software
Operating System Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10