Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Brady Workstation Asset Tracking Software Suite, Minimum RAM Required 2GB, E-Media

Catalog No. 19129122 Shop All Brady Corporation Products
Change view
Click to view available options
Brady Workstation Asset Tracking Software Suite, Minimum RAM Required 2GB, E-Media
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-129-122 Brady Workstation Asset Tracking Software Suite, Minimum RAM Required 2GB, E-Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-122 Supplier Brady™ Supplier No. BWSASTSEM

Includes

Email with Activation Code

Requires

Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB

Brady™ Workstation Asset Tracking Software Suite Easily creates asset tags or asset location labels

Features:

  • Asset Tags - Easily create the asset tags or asset location labels you need.
  • Custom Designer - Complete, customized label creation and editing.
  • Template Editor - Create templates with locked or editable elements for consistent label creation.
  • Excel Import - Easily import Excel data right onto your labels.
  • Advanced Import - Easily import data from sources beyond Excel onto your labels.
  • Advanced Sequence - Create dependent and complex sequences for label creation.

Specifications

For Use With (Equipment) Brady Label Printers
Resolution 1024 x 768
Software Type Brady Workstation Asset Tracking Software Suite
Operating System Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.