Learn More
Brady™ Brady Workstation Flip Flop, Minimum RAM Required 2GB, E-Media
Catalog No. 19129119 Shop All Brady Corporation Products
Change view
Click to view available options
Brady Workstation Flip Flop, Minimum RAM Required 2GB, E-Media
Catalog No.
19-129-119 Brady Workstation Flip Flop, Minimum RAM Required 2GB, E-Media
1 options
Catalog No. 19-129-119 Supplier Brady™ Supplier No. BWSFFEM
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Brady™ Workstation Flip Flop software lets users automatically swap text items horizontally or vertically on labels with the Flip Flop add-on module.
Features:
  • Especially useful for DataComm applications to reorder your data.
  • Flip selected or all text on the label
  • Remove or retain spacing after removing the flip character
  • Works with Custom Designer, Custom Designer Lite and Text Labels
Specifications
For Use With (Equipment) Brady Label Printers
Resolution 1024 x 768
Software Type Brady Workstation Flip Flop
Operating System Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10