Learn More
Brady™ Brady Workstation Pipemarker Software, Minimum RAM Required 2GB, E-Media
Shop All Brady Corporation ProductsDescription
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Features:
- Easily create ANSI/ASME compliant pipe markers using pre-defined templates.
- Pipe marker length set based on pipe diameter requirement
- Common chemicals pre-populated with classification
- Hundreds of graphics available, including arrows to indicate flow direction
Specifications
Specifications
| For Use With (Equipment) | Brady Label Printers |
| Languages | Bulgarian, Chinese (Simplified), Czech, Danish, German, English, Spanish, Estonian, Finnish, French, Croatian, Hungarian, Italian, Japanese, Korean, Dutch, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Swedish, Chinese (Traditional), Turkish |
| Resolution | 1024 x 768 |
| Software Type | Brady Workstation Pipemarker Software |
| Operating System | Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10 |