Learn More
Brady™ Brady Workstation Print Partner Software Suite, E-Media
Catalog No. 19129101 Shop All Brady Corporation Products
Change view
Click to view available options
Brady Workstation Print Partner Software Suite, E-Media
Catalog No.
19-129-101 Brady Workstation Print Partner Software Suite, E-Media
1 options
Catalog No. 19-129-101 Supplier Brady™ Supplier No. BWSPPSEM
Includes
Email with Activation Code
Brady™ Workstation Print Partner Suite is a package of the Brady™ Workstation Print Partner application and Advanced Import and Sequence extensions in a single bundle.
  • Includes Brady™ Workstation Print Partner, which is deployable on multiple locations where only printing is required with no design capabilities
  • Brady™ Workstation Advanced Import app which allows for importation of data from other sources
  • Brady™ Workstation Sequence app which allows for creating a series of labels
  • Locked templates ensure label data is consistent and reduces chances for user error
Specifications
Product Type Software
For Use With (Equipment) Brady Label Printers
Width (English) 0.002 in.
Includes Email with Activation Code
Software Type Brady Workstation Print Partner Software Suite
Operating System Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8 Professional (32 or 64 bit), Windows 7 Ultimate, Windows 7 Professional, Windows 7 Enterprise, Windows 10