Learn More
Brady™ Brady Workstation Print Partner Software Suite, Multi-User, CD
Catalog No. 19129103 Shop All Brady Corporation Products
Change view
Click to view available options
Brady Workstation Print Partner Software Suite, Multi-User, CD
Catalog No.
19-129-103 Brady Workstation Print Partner Software Suite, Multi-User, CD
1 options
Catalog No. 19-129-103 Supplier Brady™ Supplier No. BWSPPSCDVOL
Includes
CD
Brady™ Workstation Print Partner Suite is a package of the Brady™ Workstation Print Partner application and Advanced Import and Sequence extensions in a single bundle.
Features:
  • Includes Brady™ Workstation Print Partner which is deployable on multiple locations where only printing is required with no design capabilities
  • Brady™ Workstation Advanced Import app which allows for importation of data from other sources
  • Brady™ Workstation Sequence app which allows for creating a series of labels
  • Locked templates ensure label data is consistent and reduces chances for user error
  • Multi user version of the Print Partner Suite software
  • Minimum Order Quantity: 2
Specifications
For Use With (Equipment) Brady Label Printers
Includes CD
Software Type Brady Workstation Print Partner Software Suite
Operating System Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8 Professional (32 or 64 bit), Windows 7 Ultimate, Windows 7 Professional, Windows 7 Enterprise, Windows 10