Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Brady Workstation Product and Wire Identification Software Suite - E-Media

Catalog No. 19129113 Shop All Brady Corporation Products
Change view
Click to view available options
Brady Workstation Product and Wire Identification Software Suite - E-Media
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-129-113 Brady Workstation Product and Wire Identification Software Suite - E-Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-113 Supplier Brady™ Supplier No. BWSPWIDSEM

Includes

Email with Activation Code

Requires

Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB

Brady™ Workstation Product and Wire Identification Suite is the go-to solution for creating product identification and wire marking labels

Features:

  • Custom Designer - Complete, customized label creation and editing.
  • Text Labels - Simple designing for text-only wire and cable labels.
  • Sequence - Easily create labels based on numeric and alphanumeric sequences.
  • Excel Import - Easily import Excel data right onto your labels.
  • Advanced Import - Easily import data from sources beyond Excel onto your labels.
  • Template Editor - Create templates with locked or editable elements for consistent label creation.

Specifications

Product Type Software
For Use With (Equipment) Brady Label Printers
Width (English) 0.002 in.
Resolution 1024 x 768
Software Type Brady Workstation Product and Wire Identification Software Suite
Operating System Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.