Learn More
Brady™ Brady Workstation Safety and Facility Identification Software Suite, E-Media
Shop All Brady Corporation ProductsDescription
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Features:
- Custom Designer - Complete, customized label creation and editing.
- Express Signs - Create safety and facility signs in minutes with predefined templates and layouts.
- Arc Flash - Create NFPA compliant arc flash labels in a few clicks.
- Arrow Tape - Create arrow tape labels with predefined templates to show pipe flow direction.
- Pipemarker - Easily create ANSI/ASME compliant pipe markers using pre-defined templates.
- Sequence - Easily create labels based on numeric and alphanumeric sequences.
Specifications
Specifications
| Product Type | Software |
| For Use With (Equipment) | Brady Label Printers |
| Width (English) | 0.002 in. |
| Format | Email with Activation Code |
| Includes | Email with activation code |
| Languages | Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Spanish, Swedish, Turkish, Vietnamese |
| Resolution | 1024 x 768 |
| Software Type | Brady Workstation Safety and Facility Identification Software Suite |
| Operating System | Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10 |
| Alternate Part Number | BWS-SFIDS-NA-EM |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.