Learn More
Brady™ Brady Workstation Text Labels Software, Minimum RAM Required 2GB, E-Media
Shop All Brady Corporation ProductsDescription
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Features:
- Create and edit text-only labels.
- Works with all extensions for importing data, creating sequential labels and editing templates.
- This app fully supports opening saved “line mode” files from LabelMark 5 and 6 with no conversion required
Specifications
Specifications
| For Use With (Equipment) | Brady Label Printers |
| Languages | Bulgarian, Chinese , Czech, Danish, German, English, Spanish, Estonian, Finnish, French, Croatian, Hungarian, Italian, Japanese, Korean, Dutch, Norwegian, Mexican-Spanish, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Swedish, Tradditional Chinese, Turkish |
| Resolution | 1024 x 768 |
| Software Type | Brady Workstation Text Labels Software |
| Operating System | Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10 |