Learn More
Brady™ Brady Workstation Wire Identification Suite, Minimum RAM Required 2GB, E-Media
Shop All Brady Corporation ProductsDescription
Includes
Email with Activation Code
Requires
Internet Connection Required Yes, Free Disk Space Required: 500 MB plus 30 MB for each installed app MB
Features:
- Text Labels - Simple designing for text-only wire and cable labels.
- Template Editor - Create templates with locked or editable elements for consistent label creation.
- Flip Flop - Automatically swap text items horizontally or vertically for DataCom labeling.
- Advanced Import - Easily import data from sources beyond Excel onto your labels.
- Advanced Sequence - Create dependent and complex sequences for label creation.
- Label Settings - Manage printing adjustments and custom label supplies.
Specifications
Specifications
| For Use With (Equipment) | Brady Label Printers |
| Resolution | 1024 x 768 |
| Software Type | Brady Workstation Wire Identification Suite |
| Operating System | Windows 8.1 Professional, Windows 8.1 Enterprise, Windows 8.1 (32- or 64-bit), Windows 8 Professional (32 or 64 bit), Windows 8 (32- or 64-bit), Windows 7 Ultimate, Windows 7 SPI, Windows 7 Professional, Windows 7 Enterprise, Windows 7 (32- or 64-bit), Windows 10 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.