Learn More
MSC Brady Woven Barricade Tape, Polypropylene, 1 Roll
Catalog No. 5000685159
Encompass
This woven barricade tape, made of B-903 polypropylene with ultraviolet inhibitors, is used to mark outdoor hazards. It provides fast on-the-spot warning of potential danger with black & yellow horizontal warning stripes.
  • Made of polypropylene material
  • Features ultraviolet inhibitors for outdoor durability
  • Designed for outdoor hazard marking
  • Provides visual warning with black & yellow horizontal stripes
  • Has a 2 in width
  • Has a 200 ft length
  • Features 3 mil thickness
  • RoHS compliant
Catalog No. 50-006-85159 Supplier MSC Supplier No. 76000140
This woven barricade tape, made of B-903 polypropylene with ultraviolet inhibitors, is used to mark outdoor hazards. It provides fast on-the-spot warning of potential danger with black & yellow horizontal warning stripes.
  • Made of polypropylene material
  • Features ultraviolet inhibitors for outdoor durability
  • Designed for outdoor hazard marking
  • Provides visual warning with black & yellow horizontal stripes
  • Has a 2 in width
  • Has a 200 ft length
  • Features 3 mil thickness
  • RoHS compliant