Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Woven Barricade Tape

Catalog No. 19041057 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-041057 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-041057 Supplier Brady™ Supplier No. 91176
  • $112.00
    $83.65 / Each of 1

    Save $28.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Woven Barricade Tape is a woven barricade tape, made of B-903 polypropylene with ultraviolet inhibitors, to mark outdoor hazards.

  • Made of B-903 Woven Polypropylene from Brady™
  • Contains special ultraviolet inhibitors for permanent outdoor marking
  • Visual barricading provides fast on-the-spot warning of potential danger
  • Woven barricade tape with black and yellow horizontal warning stripes

Specifications

Certifications/Compliance OSHA 1910.144(a)
Adhesive Type Non-adhesive
Color Black, Yellow
Finish Matte
Format Tape
Material Polypropylene
Printable Sides Single-side Printable
Product Type Tape
Sterility Non-sterile
For Use With (Application) For permanent outdoor marking
Quantity 1 Ea.
Thickness (English) 3 mil
Dimensions (L x W) 200 ft. x 2 in.
Length (English) 200 ft.
Length (Metric) 60.96 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.