Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Yellow Chains and Replacement Links for Bradylink™ Warning Posts

Catalog No. 19017242 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-017242 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-017242 Supplier Brady™ Supplier No. 78234
  • $650.50
    $341.65 / Pack of 1

    Save $308.85

    47% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offers decorative alternative to metal chains

  • Made of durable polyethylene
  • Decorative alternative to metal chains
  • Color coded to match our Bradylink warning posts and have a special link design that prevents kinking
  • Standard package 100 feet

Chains are not intended to support heavy objects.

Compliance

Pass 2011/65/EU

Specifications

Color Yellow
Length (Metric) 30.48 m
For Use With (Application) Recommended for indoor use only
Includes 36 links/PK for chain
Length (English) 100 ft.
Material Polyethylene
Type Link Warning Chain

Recommended for indoor use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.