Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyGlo™ Egress Marking Tape Roll, V-Arrows, Luminous on Green

Catalog No. 19103818 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Green on Glow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-818 Green on Glow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-818 Supplier Brady™ Supplier No. 90972
  • $200.00
    $135.65 / Each of 1

    Save $64.35

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for safe low level lighting egress pathway marking during power outages and emergency situations

Photoluminescent Polyester

Specifications

Color Green on Glow
Material Phosphorescent Polyester
Width (English) 3 in.
Width (Metric) 101.6 mm
Thickness 0.008 in.
Length (English) 5 yd.
Packaging Quantity 1 Ea.
Displayed Text Blank
Type Marking Tape
Adhesive Type Self-adhesive
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.