Learn More
Brady™ Phosphorescent Polyester Laminated Polystyrene Sign: HIGH VOLTAGE - KEEP OUT/ALTO VOLTAJE - NO ACERCARSE
Shop All Brady Corporation ProductsDescription
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Specifications
| Material | Polyester, Polystyrene |
| Mounting Type | Corner Holes |
| Shape | Rectangular |
| Languages | English, Spanish |
| Dimensions (L x W) | 7 x 10 in. |
| Displayed Text | HIGH VOLTAGE - KEEP OUT/ALTO VOLTAJE - NO ACERCARSE |
| Product Type | Safety Sign |
| Color | Red on Glow |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.