Learn More
Brady™ Phosphorescent Polyester Laminated Polystyrene Sign: HIGH VOLTAGE. WILL RESULT IN SERIOUS SHOCK OR DEATH.
Catalog No. 19109573 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
Catalog No. Dimensions (L x W)
19-109-573 10 x 7 in.
1 options
Catalog No. 19-109-573 Supplier Brady™ Supplier No. 144404
Improve workplace safety with proper signage and markings. This phosphorescent Brady™ laminated polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Polyester, Polystyrene
Shape Rectangular
Dimensions (L x W) 10 x 7 in.
Displayed Text HIGH VOLTAGE. WILL RESULT IN SERIOUS SHOCK OR DEATH.
Color Black on White