Learn More
Brady™ BradyJet™ Continuous Polyester Labels
Brady™ BradyJet™ Continuous Polyester Labels are for use with BradyJet™ J2000 and are ideal for a wide range of identification applications.
Supplier:  Brady™ J20C40002569
 View more versions of this product
Catalog No. 19-129-152
Add to Cart
Description
Description
  • Constructed of indoor-use general purpose polyester
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Specifications
Acrylic, Permanent
White
Roll
Single-side Printable
Label
Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking, Lockout Tagout Labeling, GHS and Chemical Labels, Arc Flash Labels
1
1 Label/Bag
-40°C to +60°C
0.119 mm
100 ft.
4 in.
1.88 in.
None
Matte
Polyester
Ink Jet
Non-sterile
BradyJet™ J2000 Printer
-40°C
-40°F to +140°F
0.0047 in.
100 ft. x 4 in.
30.48 m
101.60 mm
J20C-4000-2569
Product Suggestions
Product Suggestions