Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyJet™ J2000 Ink Pads

Catalog No. 19129145 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-145 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-145 Supplier Brady™ Supplier No. J20PAD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brady™ BradyJet™ J2000 Ink Pads includes one bottom felt pad, one foam strip, and one top felt strip.

  • Use replacement ink pads after 20,000 prints, or signs of excessive ink saturation

Specifications

Color White
For Use With (Equipment) Brady Label Printers
Material Foam
Dimensions (L x W x H) 12 x 0.25 x 7 in.
Includes 1 bottom felt pad, 1 foam strip, 1 top felt strip
Alternate Part Number J20-PAD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.