Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyJet™ J2000 Polypropylene Labels

Catalog No. 19129284 Shop All Brady Corporation Products
Change view
Click to view available options
Size (English):
1 x 3 in.
100 ft. x 1.125 in.
100 ft. x 2.25 in.
3.35 x 2.17 in.
4.33 x 2.95 in.
6.1 x 4.13 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English)
19-129-284 100 ft. x 1.125 in.
19-129-281 1 x 3 in.
19-129-279 4.33 x 2.95 in.
19-129-280 6.1 x 4.13 in.
19-129-283 100 ft. x 2.25 in.
19-129-278 3.35 x 2.17 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-129-284 Supplier Brady™ Supplier No. J20C11257425J
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical resistant, low temperature labels ideal for full-color identification and barcode applications

  • This B-7425J polypropylene material is ideal for full-color identification and barcode applications where resistance to liquid nitrogen, autoclave conditions and harsh chemicals is required
  • Matte white polypropylene with acrylic adhesive is inkjet printable
  • Perfect for laboratory identification where labels may come in contact with harsh chemicals
  • Service temperature of -112 to 158° F (-80 to 70° C)
  • Specifications

    Autoclavable Autoclavable
    Adhesive Type Acrylic, Permanent
    Coating None
    Color White
    Finish Matte
    Format Continuous
    Material Polypropylene
    Printable Sides Single-side Printable
    Print Technology Ink Jet
    Product Type Label
    Sterility Non-sterile
    For Use With (Application) Barcode labelling, Cold temperature labelling, Component and equipment labelling, General identification, Hazard communication identification, Laboratory labelling
    For Use With (Equipment) BradyJet™ J2000
    Labels per Row 1
    Min. Operating Temperature (Metric) -80°C
    No. per Pack 1 Tape/Bag
    Quantity 1 Ea.
    Size (English) 100 ft. x 1.125 in.
    Temperature Range (English) -112°F to +158°F
    Temperature Range (Metric) -80°C to +70°C
    Thickness (English) 0.005 in.
    Thickness (Metric) 0.127 mm
    Dimensions (L x W) 100 ft. x 1.125 in.
    Length (English) 100 ft.
    Length (Metric) 30.48 m
    Width (English) 1.5 in.
    Width (Metric) 28.58 mm
    Alternate Part Number J20C-1125-7425J
    Show More Show Less
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.