Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyJet™ J4000 Chemical-resistant Continuous Polypropylene Labels

Catalog No. p-7238944 Shop All Brady Corporation Products
Change view
Click to view available options
Width (English):
2.25 in.
4 in.
6 in.
8 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
17-001-032 2.25 in.
17-001-033 4 in.
17-001-034 6 in.
17-001-035 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-001-032 Supplier Brady™ Supplier No. J40C22507425J
Only null left
Add to Cart
Add to Cart

This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.

The B-7425J polypropylene material of these labels is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required. An ink cartridge is required to print this material.

  • Matte polypropylene with acrylic adhesive is printable with Inkjet
  • Low-cost, full-color GHS labeling option withstands chemical exposure
  • Service temperature of -112 to 158°F (-80 to 70°C)

Specifications

Autoclavable Autoclavable
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Continuous
Material Polypropylene (B-7425J)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification, Component and Equipment Labeling, General Identification
For Use With (Equipment) Brady Label Printers
Labels per Row 1 Label
Temperature Range (English) -112°F to 158°F
Thickness (English) 0.005 in.
Length (English) 100 ft.
Width (English) 2.25 in.
Width (Metric) 57.15 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.