Learn More
CPC Scientific H-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific BRAK001A
Encompass
Bradykinin that contracts smooth muscles, relaxes blood vessels and increases vascular permeability. Additional physiological functions include the stimulation of pain receptors, inhibition of cAMP accumulation, and induction of the release of nitric oxide. ONE-LETTER SEQUENCE: RPPGFSPFRMOLECULAR FORMULA: C50H73N15O11MOLECULAR WEIGHT:1060.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1490132], RESEARCH AREA: CardiovascularREFERENCES: D. Regoli et al., J. Pharmacol. Rev., 32, 1 (1980)
Catalog No. 50-210-8845