Learn More
Medchemexpress LLC HY-P1488 1mg Medchemexpress, Bradykinin (1-5) CAS:23815-89-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP14881MG
Encompass_Preferred
Medchemexpress, HY-P1488 1mg Bradykinin (1-5) CAS:23815-89-6 Bradykinin (1-5) is a major stable metabolite of Bradykinin, formed by the proteolytic action of angiotensin-converting enzyme (ACE). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3060