Learn More
Medchemexpress LLC HY-P1469 5mg Medchemexpress, Bradykinin (1-6) CAS:23815-88-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP14695MG
Encompass_Preferred
Medchemexpress, HY-P1469 5mg Bradykinin (1-6) CAS:23815-88-5 Bradykinin (1-6) is an amino-truncated Bradykinin peptide. Bradykinin (1-6) is a stable metabolite of Bradykinin, cleaved by carboxypeptidase Y (CPY). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3055