Learn More
Medchemexpress LLC HY-P1490 5mg Medchemexpress, Bradykinin (2-9) CAS:16875-11-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP14905MG
Encompass_Preferred
Medchemexpress, HY-P1490 5mg Bradykinin (2-9) CAS:16875-11-9 Bradykinin (2-9) is an amino-truncated Bradykinin peptide. Bradykinin (2-9) is a metabolite of Bradykinin, cleaved by Aminopeptidase P. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0535