Learn More
Enzo Life Sciences Bradykinin (25mg). CAS: 5979-11-3
SDP
Supplier:  Enzo Life Sciences ALX152006M025
Encompass_Preferred
eNOS activator. Activator of eNOS (NOS III). Formula: C50H73N15O11. MW: 1060.3. Purity: ≥98% (HPLC). Appearance: White to off-white solid. MI: 14: 1359. Solubility: Soluble in 5% acetic acid. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-7785