Learn More
Eurofins Discoverx Bradykinin, 5 mg. (92-1053)
Supplier:  Eurofins Discoverx 921053
Encompass
Bradykinin is a peptide that acts as a mediator of pain and inflammation, with pro-inflammatory activity. It is an agonist of the bradykinin receptors, with selectivity for B2 receptor over B1. Important roles in fluid regulation, electrolyte balance, smooth muscle contraction, vasodilation and capillary permeability. A common target of blood pressure lowering drugs, increased levels of bradykinin may be associated with increased lung and gastric cancer risk. Provided as 5 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9594