Learn More
Medchemexpress LLC HY-P0206 5mg Medchemexpress, Bradykinin CAS:58-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02065MG
Encompass_Preferred
Medchemexpress, HY-P0206 5mg Bradykinin CAS:58-82-2 Bradykinin is an active peptide that is generated by the kallikrein-kinin system. It is a inflammatory mediator and also recognized as a neuromediator and regulator of several vascular and renal functions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4050