Learn More
Apexbio Technology LLC Bradykinin (acetate) 6846-03-3 10mg
SDP
Catalog No. 5000091449
Encompass_Preferred
Bradykinin acetate (CAS 6846-03-3) is a peptide agonist targeting G protein-coupled B1 and B2 receptors It is designed to activate bradykinin receptors thereby modulating inflammatory responses vascular tone and endothelial signaling pathways Bradykinin acetate exerts its biological activity primarily through endothelium-dependent release of nitric oxide and prostacyclin leading to vasodilation Receptor activation also triggers non-vascular smooth muscle contraction increases vascular permeability and mediates pain signaling pathways Based on these pharmacological properties Bradykinin acetate holds research potential in studies of inflammation pain mechanisms and endothelial cell signaling
Catalog No. 50-000-91449 Supplier Apexbio Technology LLC Supplier No. C412610
Bradykinin acetate (CAS 6846-03-3) is a peptide agonist targeting G protein-coupled B1 and B2 receptors It is designed to activate bradykinin receptors thereby modulating inflammatory responses vascular tone and endothelial signaling pathways Bradykinin acetate exerts its biological activity primarily through endothelium-dependent release of nitric oxide and prostacyclin leading to vasodilation Receptor activation also triggers non-vascular smooth muscle contraction increases vascular permeability and mediates pain signaling pathways Based on these pharmacological properties Bradykinin acetate holds research potential in studies of inflammation pain mechanisms and endothelial cell signaling