Learn More
Medchemexpress LLC Bradykinin acetate | 6846-03-3 | 99.58% | 1060.21 (free base) | 5 MG
SDP
Catalog No. 5000425362
Encompass_Preferred
Bradykinin acetate is an effective endothelium-dependent vasodilator that can lower blood pressure. It induces contraction of bronchial and intestinal non-vascular smooth muscle, increases vascular permeability, and participates in pain mechanisms. It is a potent vasodilator peptide that stimulates specific endothelial B2 receptors, causing the release of prostacyclin, NO, and EDHF.
  • Effective endothelium-dependent vasodilator
  • Lowers blood pressure
  • Induces contraction of bronchial and intestinal non-vascular smooth muscle
  • Increases vascular permeability
  • Participates in pain mechanisms
  • Stimulates specific endothelial B2 receptors
  • Causes release of prostacyclin, NO, and EDHF
Catalog No. 50-004-25362 Supplier Medchemexpress LLC Supplier No. HYP0206A5MG
Bradykinin acetate is an effective endothelium-dependent vasodilator that can lower blood pressure. It induces contraction of bronchial and intestinal non-vascular smooth muscle, increases vascular permeability, and participates in pain mechanisms. It is a potent vasodilator peptide that stimulates specific endothelial B2 receptors, causing the release of prostacyclin, NO, and EDHF.
  • Effective endothelium-dependent vasodilator
  • Lowers blood pressure
  • Induces contraction of bronchial and intestinal non-vascular smooth muscle
  • Increases vascular permeability
  • Participates in pain mechanisms
  • Stimulates specific endothelial B2 receptors
  • Causes release of prostacyclin, NO, and EDHF