Learn More
Medchemexpress LLC Bradykinin acetate | 6846-03-3 | 99.6% | 1060.21 | 10 MG
SDP
Catalog No. 5000425885
Encompass_Preferred
Bradykinin acetate is a potent endothelium-dependent vasodilator known for its ability to lower blood pressure. It is involved in inducing contraction of bronchial and intestinal non-vascular smooth muscle, increasing vascular permeability, and participating in pain mechanisms. This compound is suitable for various research applications, including studies on its role in cardiovascular function, inflammation, and cellular signaling pathways.
  • Effective endothelium-dependent vasodilator
  • Lowers blood pressure
  • Induces contraction of bronchial and intestinal non-vascular smooth muscle
  • Increases vascular permeability
  • Participates in pain mechanisms
  • Stable for 2 years at -80°C as powder
  • Stable for 6 months at -80°C in solvent under sealed, moisture-free conditions
Catalog No. 50-004-25885 Supplier Medchemexpress LLC Supplier No. HYP0206A10MG
Bradykinin acetate is a potent endothelium-dependent vasodilator known for its ability to lower blood pressure. It is involved in inducing contraction of bronchial and intestinal non-vascular smooth muscle, increasing vascular permeability, and participating in pain mechanisms. This compound is suitable for various research applications, including studies on its role in cardiovascular function, inflammation, and cellular signaling pathways.
  • Effective endothelium-dependent vasodilator
  • Lowers blood pressure
  • Induces contraction of bronchial and intestinal non-vascular smooth muscle
  • Increases vascular permeability
  • Participates in pain mechanisms
  • Stable for 2 years at -80°C as powder
  • Stable for 6 months at -80°C in solvent under sealed, moisture-free conditions