Learn More
Enzo Life Sciences Bradykinin ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900206
Encompass_Preferred
Sensitive and reliable ELISA for measuring bradykinin in plasma, serum and urine samples. The Bradykinin EIA kit is a colorimetric, competitive immunoassay kit with results in 3 hours. Sensitivity: 24.8 pg/ml (range 11.7-30,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter Plate, Conjugate, Antibody, Assay Buffer 16, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Streptavidin-HRP
Catalog No. NC1533235