Learn More
Abcam Bradykinin ELISA kit, 1 x 96 Tests
Supplier:  Abcam AB1369361X96TESTS
Encompass_Preferred
Bradykinin ELISA kit ab136936 is used to quantify Bradykinin with a sensitivity of 55 pg/ml. Competitive ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0827768