Learn More
Cayman Chemical Bradykinin (human, mouse, rat, bovine) (acetate), Purity: ≥98%, CAS: 6846-03-3, 10 mg
SDP
Catalog No. 502916172
Encompass_Preferred
An endogenous vasodilator; binds to the bradykinin B2 receptor (IC50 = 0.54 nM in COS-7 cells expressing the human receptor) and acts as an agonist but does not bind the bradykinin B1 receptors (Ki = >10,000 nM in HEK293 cells expressing the human receptor); stimulates arachidonic acid release in CHO cells expressing the human bradykinin B2 receptor (EC50 = 0.7 nM); induces relaxation of ACh- or PDBu precontracted isolated porcine coronary arterial rings at 100 nM; induces hyperalgesia in mice at 1 µg/paw; plasma levels are increased in patients during an acute attack of angioedema
Catalog No. 50-291-6172 Supplier Cayman Chemical Supplier No. 3740810 MG
An endogenous vasodilator; binds to the bradykinin B2 receptor (IC50 = 0.54 nM in COS-7 cells expressing the human receptor) and acts as an agonist but does not bind the bradykinin B1 receptors (Ki = >10,000 nM in HEK293 cells expressing the human receptor); stimulates arachidonic acid release in CHO cells expressing the human bradykinin B2 receptor (EC50 = 0.7 nM); induces relaxation of ACh- or PDBu precontracted isolated porcine coronary arterial rings at 100 nM; induces hyperalgesia in mice at 1 µg/paw; plasma levels are increased in patients during an acute attack of angioedema