Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bradylink Warning Post System Accessory - Stanchion Chain, Black, 2 in.

Catalog No. 19104387 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-387 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-387 Supplier Brady™ Supplier No. 78268
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bradylink Warning Post Chains are made of colorful polyethylene and are a decorative, rust-free alternative to metal stanchion chains.

  • Chains are color coded to match our Bradylink Warning Posts and have a special link design that prevents kinking
  • Bradylink Chains are not intended to support heavy objects and are recommended for indoor use only
  • Package includes chain only (stanchions sold separately)
  • The Bradylink Warning Post System is non-conductive so it can be safely used in electrically hazardous areas

Specifications

Color Black
Width (English) 2 in.
Width (Metric) 290.83 mm
Packaging Quantity 1 Ea.
Length (English) 100 ft.
Type Warning Chains
Dimensions (L x W) 100 x 2 ft.
Material Polyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.