Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Bradylink Warning Stanchion - Non-Reflective Stripes

Catalog No. 19104352 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-352 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-352 Supplier Brady™ Supplier No. 80940

missing translation for 'validMainframeMsg'

Bradylink Warning Posts are made of colorful polystyrene and polyethylene and are a decorative, rust-free alternative to metal posts. Each post includes base, top and c-hook.

  • Posts are non-conductive so they can be safely used in electrically hazardous areas
  • Hollow base can be filled with sand for more stable application
  • Bradylink Chains sold separately
  • Non-reflective stripes alternate every 4 in. for greater visibility

Specifications

Color Black, Yellow
Dimensions (Dia. x H) 2.5 x 40 in.
Width (Metric) 408.94 mm
Height (English) 40 in.
Packaging Quantity 1 Ea.
Material Polyethylene, Polystyrene
Diameter (English) Base 14 in.
Type Stanchion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.