Learn More
Brady BradyPrinter i3300
Catalog No. 17033764 Shop All Brady Corporation Products
Change view
Click to view available options
Software Type:
PWID Software
SFID Software
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Software Type
17-033-764 PWID Software
17-033-777 SFID Software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-033-764 Supplier Brady Supplier No. 177531
Add to Cart
Add to Cart
Seamlessly connect the Brady products you rely on with a printer and scanner system.
  • Get auto setup, auto calibration and fast changeovers with the i3300 printer.
  • Enjoy seamless, LabelSense™ technology that instantly recognizes label materials and ribbons
  • Create labels with text or graphics with the included label creation software
Specifications
Product Type Label Printer Kit
Color Blue, Gray
Display Type LCD Touchscreen
For Use With (Equipment) B30 Series Thermal Transfer Tapes, Brady Workstation, B30 Series Thermal Transfer Tapes and Ribbons, Brady Labels, B30 Series Thermal Transfer Tapes
Material Plastic
Print Technology Thermal Transfer
Length (English) 12 in.
Width (English) 9 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Includes V4500 Barcode Scanner
Model i3300
Resolution 300 DPI
Software Type PWID Software
Voltage 115/230 VAC