Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyPrinter i3300-C Sign and Label Printer WIFI

Catalog No. 17607628 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Blue, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-607-628 Blue, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-607-628 Supplier Brady™ Supplier No. 149552
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Get your ideal label printing scenario with the BradyPrinter i3300 Industrial Label Printer, labels and ribbons that snap into place, no calibration and no wasted blank labels.

Get your ideal label printing scenario with the BradyPrinter i3300 Industrial Label Printer, labels and ribbons that snap into place, no calibration and no wasted blank labels.

  • Mid-volume industrial label printer ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels.
  • Print smart technology allows the printer to print on the first label, even sleeves and small labels.
  • Changeovers in a flash, foolproof drop-in label rolls and ribbons snap into place in 20 seconds.
  • Talk about options, this printer prints on more than 825 parts and 70 material options for facility-wide identification.
  • Add one of the Brady Workstation Software Suites to your solution and make label creation even easier.
  • Setup is stress free: Contact our Brady Technical Support Team and we will schedule a free phone setup appointment.

Specifications

Product Type Industrial Label Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) B30 Series Thermal Transfer Tapes and Ribbons
Material Plastic
Print Technology Thermal Transfer
Length (English) 12 in.
Width (English) 9 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.