Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady BradyPrinter i5300

Catalog No. 17033776 Shop All Brady Corporation Products
Change view
Click to view available options
Software Type:
PWID Software
SFID Software
Resolution:
300 DPI
600 DPI
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Software Type Resolution
17-033-776 SFID Software 600 DPI
17-033-765 PWID Software 300 DPI
17-033-775 SFID Software 300 DPI
17-033-766 PWID Software 600 DPI
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-033-776 Supplier Brady Supplier No. 177543
Add to Cart
Add to Cart

Seamlessly connect the Brady products you rely on with a printer and scanner system.

  • Use the i5300 for fast and efficient printing with zero setup, zero adjustment and zero wasted labels.
  • Create labels with text or graphics with the included label creation software

Specifications

Product Type Label Printer Kit
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) B30 Series Thermal Transfer Tapes, Brady Labels, Brady Workstation
Material Plastic
Print Technology Direct Thermal, Thermal Transfer
Length (English) 15.8 in.
Width (English) 11.2 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Includes V4500 Barcode Scanner
Resolution 600 DPI
Software Type SFID Software
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.