Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BradyPrinter M611 Magnet

Catalog No. 19129179 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-179 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-129-179 Supplier Brady™ Supplier No. BMPMAGNET2
This item is not returnable. View return policy

Brady™ BradyPrinter M611 Magnet gives the convenient option to attach your label printer to panels, I-beams and any other magnetic-receptive surface during use or storage.

  • Designed for use with the BradyPrinter M611 Mobile Printer
  • Allows printer to affix to panel door or metal surface for more hands-free operation or storage
  • Attaches to back of printer with screws to ensure stability

Specifications

Color Black
For Use With (Equipment) BradyPrinter M611
Diameter (English) 4.38 in.
Diameter (Metric) 11.13 cm
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.