Learn More
Brady™ BradyPrinter™ PR360 Plus Printer with LabelMark™ Label Design Software
Catalog No. 19104491 Shop All Brady Corporation Products
Change view
Click to view available options
Bradyprinter™ PR360 Plus Printer with LabelMark™ Label Design Software
Catalog No.
19-104-491 Bradyprinter™ PR360 Plus Printer with LabelMark™ Label Design Software
1 options
Catalog No. 19-104-491 Supplier Brady™ Supplier No. BPPR360+LMS
BradyPrinter™ PR360+ with LabelMark™ Standard Software offers high performance THT printing for cable ID applications, versatile label design, and prints labels up to 6 in. wide.
  • BradyPrinter™ PR360+ printer is perfect for use where material has a long life expectancy
  • LabelMark™ Standard Software takes the hassle out of any labeling project from basic label design and printing to advanced import and editing options
  • Designed for 300 dpi printing on labels up to 6 in. wide in volumes of up to 7000 labels per day at 12hrs / 5 days per week operation
  • Perfect for data communications labeling, general & industrial labeling, wire & cable marking, barcode labeling, and more
Specifications
Product Type Printer Kit
Autoclavable Not Autoclavable
Description Printer Kit
Includes BradyPrinter™ PR+ 360 Printer
Quantity Kit