Learn More
Brady™ BradyPrinter™ PR600 Plus Printer with LabelMark™ Label Design Software
Catalog No. 19104492 Shop All Brady Corporation Products
Change view
Click to view available options
Bradyprinter™ PR600 Plus Printer with LabelMark™ Label Design Software
Catalog No.
19-104-492 Bradyprinter™ PR600 Plus Printer with LabelMark™ Label Design Software
1 options
Catalog No. 19-104-492 Supplier Brady™ Supplier No. BPPR600+LMS
BradyPrinter™ PR300+ with LabelMark™ Standard Software offers high performance, 600 dpi THT printing and label design for your cable identification needs.
  • BradyPrinter™ PR600+ printer is perfect for use where material has a long life expectancy
  • LabelMark™ Standard Software takes the hassle out of any labeling project from basic label design and printing to advanced import and editing options
  • Designed for 600 dpi printing on labels up to 4 in. wide in volumes of up to 7000 labels per day at 12hrs / 5 days per week operation
  • Perfect for data communications labeling, general & industrial labeling, wire & cable marking, barcode labeling, and more
Specifications
Product Type Printer Kit
Autoclavable Not Autoclavable
Description Printer Kit
Includes BradyPrinter™ PR+ 600 Printer
Resolution 240 x 160
Quantity Kit