Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady BradyPrinter S3100

Catalog No. 17033778 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Label Printer Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-033-778 Label Printer Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-033-778 Supplier Brady Supplier No. 177545
Add to Cart
Add to Cart

Seamlessly connect the Brady products you rely on with a printer and scanner system.

  • Use the S3100 to create the labels you need, when you need them with do-it-yourself label creation.
  • Printer is ideal for multi-user areas in your facility; anyone can simply walk up, create labels and print right on the spot
  • Label creation is even faster with the quick setup and print speeds that come with LabelSense™ technology from Brady

Specifications

Product Type Label Printer Kit
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) B30 Series Thermal Transfer Tapes, Brady Labels, Brady Workstation
Material Plastic
Print Technology Thermal Transfer
Length (English) 10.25 in,
Width (English) 12 in.
Autoclavable Not Autoclavable
Sterility Non-sterile
Includes V4500 Barcode Scanner
Resolution 300 DPI
Software Type SFID Software
Voltage 110/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.