Learn More
Carnabio Usa Inc BRAF/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0912210
Encompass
BRAF, Catalytic domain, Wild type, Amino Acid 433-726, NP_004324.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0697