Learn More
Medchemexpress LLC BRAF protein, human (active, Sf9, GST, His) | 10UG
SDP
Catalog No. 5000174052
Encompass_Preferred
Recombinant active human BRAF kinase produced in Sf9 insect cells and supplied as a GST- and His-tagged construct (residues E433-R726, UniProt P15056). Supplied as a 0.22 μm filtered solution at 1 mg/mL and validated for in vitro kinase assays and biochemical studies. Store at -80°C and avoid repeated freeze-thaw cycles to preserve activity.
  • Expressed in Sf9 insect cells for eukaryotic folding and post-translational modifications.
  • Activity validated by luminescence-based kinase assay for reliable functional studies.
  • Dual affinity tags facilitate purification and detection in assays.
  • Construct corresponds to the active kinase domain for MAPK pathway research.
  • 0.22 μm filtered, concentrated formulation suitable for biochemical assays.
  • Recommended long-term storage at -80°C to maintain activity.
Catalog No. 50-001-74052 Supplier Medchemexpress LLC Supplier No. HYP70292110UG
Recombinant active human BRAF kinase produced in Sf9 insect cells and supplied as a GST- and His-tagged construct (residues E433-R726, UniProt P15056). Supplied as a 0.22 μm filtered solution at 1 mg/mL and validated for in vitro kinase assays and biochemical studies. Store at -80°C and avoid repeated freeze-thaw cycles to preserve activity.
  • Expressed in Sf9 insect cells for eukaryotic folding and post-translational modifications.
  • Activity validated by luminescence-based kinase assay for reliable functional studies.
  • Dual affinity tags facilitate purification and detection in assays.
  • Construct corresponds to the active kinase domain for MAPK pathway research.
  • 0.22 μm filtered, concentrated formulation suitable for biochemical assays.
  • Recommended long-term storage at -80°C to maintain activity.