Learn More
Medchemexpress LLC HY-10247 10mg Medchemexpress, BRAF inhibitor CAS:918505-61-0 Purity:>98%
SDP
Catalog No. 501967988
Encompass_Preferred
Medchemexpress, HY-10247 10mg BRAF inhibitor CAS:918505-61-0 BRAF inhibitor is a B-Raf inhibitor extracted from patent WO/2011103196 A1, Compound P-0850. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7988 Supplier Medchemexpress LLC Supplier No. HY1024710MG
Medchemexpress, HY-10247 10mg BRAF inhibitor CAS:918505-61-0 BRAF inhibitor is a B-Raf inhibitor extracted from patent WO/2011103196 A1, Compound P-0850. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.