Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRAF inhibitor | 918505-61-0 | MFCD16659048 | 99.0% | 456.47 | C22H18F2N4O3S | 1 ML
SDP

Catalog No. 5000247163
Encompass_Preferred

BRAF inhibitor (Compound P-0850) is a small-molecule B-Raf kinase inhibitor with reported anti-tumor activity. It is supplied for research use as a solid and as a pre-dosed 10 mM solution in DMSO; formulation protocols, solubility data, and storage recommendations are provided for both in vitro and in vivo applications.

  • High purity (~99%).
  • Available as 10 mM solution in DMSO or as solid in multiple pack sizes.
  • Soluble in DMSO; in vitro solubility reported up to 50 mg/mL (≈109.5 mM).
  • In vivo formulation protocols provided for common vehicles (PEG300, Tween-80, SBE-β-CD, corn oil).
  • Storage recommendations given for powder and solution to preserve stability.

Catalog No. 50-002-47163 Supplier Medchemexpress LLC Supplier No. HY1024710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRAF inhibitor (Compound P-0850) is a small-molecule B-Raf kinase inhibitor with reported anti-tumor activity. It is supplied for research use as a solid and as a pre-dosed 10 mM solution in DMSO; formulation protocols, solubility data, and storage recommendations are provided for both in vitro and in vivo applications.

  • High purity (~99%).
  • Available as 10 mM solution in DMSO or as solid in multiple pack sizes.
  • Soluble in DMSO; in vitro solubility reported up to 50 mg/mL (≈109.5 mM).
  • In vivo formulation protocols provided for common vehicles (PEG300, Tween-80, SBE-β-CD, corn oil).
  • Storage recommendations given for powder and solution to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.