Learn More
Cayman Chemical B-RAF Inhibitor 1 (hydrochloride), Purity: ≥98%, CAS: 1191385-19-9, 10 mg
SDP
Catalog No. 502936728
Encompass_Preferred
An inhibitor of Raf kinases (Kis = 0.3, 1, and 1 nM for C-RAF, B-RAF, and B-RAFV600E, respectively); selective for B-RAFV600E kinase domain over Lck, Tie2, KDR, and p38α (IC50s = 83, 120, 1,000, and >1,600 nM, respectively); inhibits phosphorylation of ERK in A375 melanoma cells (IC50 = 1.8 nM); reduces tumor growth in an A375 SQ2 mouse xenograft model (ED50 = 1.3 mg/kg per day) and induces 85% tumor regression at 5 mg/kg per day for 14 days; increases tumor growth in a MIA PaCa-2 mouse xenograft model at 5 and 10 mg/kg per day
Catalog No. 50-293-6728 Supplier Cayman Chemical Supplier No. 2679210 MG
An inhibitor of Raf kinases (Kis = 0.3, 1, and 1 nM for C-RAF, B-RAF, and B-RAFV600E, respectively); selective for B-RAFV600E kinase domain over Lck, Tie2, KDR, and p38α (IC50s = 83, 120, 1,000, and >1,600 nM, respectively); inhibits phosphorylation of ERK in A375 melanoma cells (IC50 = 1.8 nM); reduces tumor growth in an A375 SQ2 mouse xenograft model (ED50 = 1.3 mg/kg per day) and induces 85% tumor regression at 5 mg/kg per day for 14 days; increases tumor growth in a MIA PaCa-2 mouse xenograft model at 5 and 10 mg/kg per day